Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500018872.mm.1 |
TC0500002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018872.mm.1 |
NM_001002897.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018872.mm.1 |
NM_001002897.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018867.mm.1 |
TC0500002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018867.mm.1 |
NM_001002897.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018867.mm.1 |
NM_001002897.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018873.mm.1 |
TC0500002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018873.mm.1 |
NM_001002897.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018873.mm.1 |
NM_001002897.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018865.mm.1 |
TC0500002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018865.mm.1 |
NM_001002897.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018865.mm.1 |
NM_001002897.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018869.mm.1 |
TC0500002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018869.mm.1 |
NM_001002897.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018869.mm.1 |
NM_001002897.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018874.mm.1 |
TC0500002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018874.mm.1 |
NM_001002897.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018874.mm.1 |
NM_001002897.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018866.mm.1 |
TC0500002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018866.mm.1 |
NM_001002897.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018866.mm.1 |
NM_001002897.3 |
Refseq |