Assay Details
| Gene Symbol: Rnf19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013923.2 | NP_038951.1 | 5 - 6 | 1513 | 86 | |
| GenBank mRNA | AF120206.1 | - | 1406 | 86 | ||
| AF120207.1 | - | 4 - 5 | 1406 | 86 | ||
| AK082665.1 | - | 5 - 6 | 1465 | 86 | ||
| AK147768.1 | - | 5 - 6 | 1501 | 86 | ||
| BC040769.1 | - | 4 - 5 | 1288 | 4457807 | 86 | |
| X71642.1 | - | 1 - 2 | 60 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rnf19a
Genomic Map