Assay Details
| Gene Symbol: Psd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289602.1 | NP_001276531.1 | 9 - 10 | 1829 | 68 | |
| NM_028707.4 | NP_082983.3 | 9 - 10 | 1826 | 68 | ||
| NR_110349.1 | 9 - 10 | 1826 | 68 | |||
| GenBank mRNA | AK018116.1 | - | 4 - 5 | 282 | 68 | |
| AK034575.1 | - | 9 - 10 | 1595 | 68 | ||
| AK045529.1 | - | 9 - 10 | 1557 | 68 | ||
| AK053835.1 | - | 9 - 10 | 1554 | 68 | ||
| AK149027.1 | - | 9 - 10 | 1556 | 68 | ||
| BC056437.1 | - | 5 - 6 | 325 | 68 | ||
| BC062930.1 | - | 8 - 9 | 1407 | 6855293 | 68 | |
| BC065053.1 | - | 9 - 10 | 1594 | 5709231 | 68 | |
| BC065055.1 | - | 9 - 10 | 1577 | 5720949 | 68 | |
| BC066026.1 | - | 9 - 10 | 1826 | 5721455 | 68 | |
| BC066036.1 | - | 9 - 10 | 1592 | 6837266 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psd2
Genomic Map