Assay Details
Gene Symbol: Nrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167891.1 | NP_001161363.1 | 3 - 4 | 1029 | 52 | |
GenBank mRNA | AW476657.1 | - | 1 - 2 | 69 | 52 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BI412864.1 | - | Chr. - - [-] on Build | 52 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
epidermal growth factor receptor signaling pathway
peptidyl-tyrosine phosphorylation
intracellular signal transduction
animal organ development
epidermal growth factor-activated receptor activity
receptor binding
epidermal growth factor receptor binding
ErbB-3 class receptor binding
epidermal growth factor receptor signaling pathway
peptidyl-tyrosine phosphorylation
intracellular signal transduction
animal organ development
epidermal growth factor-activated receptor activity
receptor binding
epidermal growth factor receptor binding
ErbB-3 class receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991821 | NM_001167891.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991821 | AW476657.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094034 | NM_001167891.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140720 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991821 | NM_001167891.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991821 | AW476657.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094034 | NM_001167891.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140720 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140720 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621622 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621636 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621589 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621609 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621689 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621659 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13966074 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621567 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621639 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13856019 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13686686 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13966073 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621660 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621686 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14068211 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621590 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621555 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14129200 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13966063 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621581 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621605 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13689712 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13850517 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621676 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621592 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13689709 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13606723 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621663 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621633 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13850497 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13689713 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13850499 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621614 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621583 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621603 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621598 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14021898 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621571 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621683 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13925581 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13925569 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621576 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621637 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13689710 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621534 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621631 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621538 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621666 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621664 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621613 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621577 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14009855 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621564 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13827731 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13689711 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621629 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621595 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621653 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621574 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621585 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621535 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13827716 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621606 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621627 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621593 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621638 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13925565 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621679 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13686688 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13984434 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621674 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621579 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621562 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14021900 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621672 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621671 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13606725 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621655 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621696 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621678 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621608 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13686696 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621668 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13966072 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621568 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13966066 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13927676 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621644 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621558 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31621560 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10458329 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1447654_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1457283_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1444491_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10458323 | - | - |
Mouse Genome 430 2.0 Array | 1444491_at | - | - |
Mouse Genome 430 2.0 Array | 1447654_at | - | - |
Mouse Genome 430 2.0 Array | 1444491_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1447654_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100042150_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100042150_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1800008745.mm.1 | NM_001167891.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800008745.mm.1 | NM_001167891.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800008745.mm.1 | TC1800001186.mm.1 | MTA TC |
Mouse Expression Set 430 | 1447654_at | - | - |
Mouse Expression Set 430 | 1444491_PM_at | - | - |
Mouse Expression Set 430 | 1444491_at | - | - |
Mouse Expression Set 430 | 1447654_PM_at | - | - |
Mouse Expression Set 430 | 1457283_PM_at | - | - |
Mouse Expression Set 430 | 1457283_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1447654_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1444491_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1447654_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1444491_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 358489 | - | - |
