Assay Details
Gene Symbol: Atp6v1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025494.3 | NP_079770.2 | 9 - 10 | 945 | 136 | |
GenBank mRNA | AB088407.1 | - | 9 - 10 | 892 | 136 | |
AK006323.1 | - | 9 - 10 | 896 | 136 | ||
AK075779.1 | - | 9 - 10 | 917 | 136 | ||
AK129007.1 | - | 7 - 8 | 2467 | 136 | ||
AK151237.1 | - | 9 - 10 | 906 | 136 | ||
AK168914.1 | - | 9 - 10 | 896 | 136 | ||
BC010217.1 | - | 9 - 10 | 893 | 3498388 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
