Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800001155.mm.1 |
NM_019706.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800001155.mm.1 |
NM_207623.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800001155.mm.1 |
TC1800000163.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800001160.mm.1 |
NM_019706.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800001160.mm.1 |
NM_207623.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800001160.mm.1 |
TC1800000163.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800001156.mm.1 |
NM_019706.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800001156.mm.1 |
NM_207623.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800001156.mm.1 |
TC1800000163.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800001161.mm.1 |
NM_019706.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800001161.mm.1 |
NM_207623.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800001161.mm.1 |
TC1800000163.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800001162.mm.1 |
NM_019706.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800001162.mm.1 |
NM_207623.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800001162.mm.1 |
TC1800000163.mm.1 |
MTA TC |