Assay Details
| Gene Symbol: Rnf138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004238.1 | - | 6 - 7 | 773 | 148 | |
| AK010486.1 | - | 6 - 7 | 762 | 148 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rnf138
Genomic Map