Assay Details
Gene Symbol: Ints4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027256.2 | NP_081532.1 | 2 - 3 | 396 | 107 | |
GenBank mRNA | AK088635.1 | - | 2 - 3 | 340 | 107 | |
AK163383.1 | - | 2 - 3 | 350 | 107 | ||
BC013710.1 | - | 2 - 3 | 342 | 3156446 | 107 | |
CA450656.1 | - | 2 - 3 | 266 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK163383.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
