Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700011476.mm.1 |
TC0700001354.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700011476.mm.1 |
NM_027256.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011476.mm.1 |
NM_027256.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011478.mm.1 |
TC0700001354.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700011478.mm.1 |
NM_027256.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011478.mm.1 |
NM_027256.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011475.mm.1 |
TC0700001354.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700011475.mm.1 |
NM_027256.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011475.mm.1 |
NM_027256.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011474.mm.1 |
TC0700001354.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700011474.mm.1 |
NM_027256.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011474.mm.1 |
NM_027256.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011477.mm.1 |
TC0700001354.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700011477.mm.1 |
NM_027256.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700011477.mm.1 |
NM_027256.2 |
Refseq |