Assay Details
| Gene Symbol: Ik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011879.2 | NP_036009.1 | 10 - 11 | 1023 | 66 | |
| GenBank mRNA | AJ006130.1 | - | 10 - 11 | 906 | 66 | |
| AK029644.1 | - | 10 - 11 | 977 | 66 | ||
| AK050625.1 | - | 10 - 11 | 981 | 66 | ||
| AK082356.1 | - | 10 - 11 | 955 | 66 | ||
| AK087976.1 | - | 10 - 11 | 969 | 66 | ||
| BC006706.1 | - | 8 - 9 | 1381 | 66 | ||
| BC014739.1 | - | 10 - 11 | 954 | 4920184 | 66 | |
| BC026569.1 | - | 10 - 11 | 954 | 4982612 | 66 | |
| BC029671.1 | - | 10 - 11 | 966 | 5364953 | 66 | |
| BC029694.1 | - | 10 - 11 | 963 | 5369477 | 66 | |
| BC033356.1 | - | 10 - 11 | 950 | 5366230 | 66 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ik
Genomic Map