Assay Details
| Gene Symbol: Ik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011879.2 | NP_036009.1 | 2 - 3 | 201 | 108 | |
| GenBank mRNA | AJ006130.1 | - | 2 - 3 | 84 | 108 | |
| AK029644.1 | - | 2 - 3 | 155 | 108 | ||
| AK050625.1 | - | 2 - 3 | 159 | 108 | ||
| AK082356.1 | - | 2 - 3 | 133 | 108 | ||
| AK087976.1 | - | 2 - 3 | 147 | 108 | ||
| AK088908.1 | - | 2 - 3 | 136 | 108 | ||
| AK163766.1 | - | 2 - 3 | 155 | 108 | ||
| AK167238.1 | - | 2 - 3 | 166 | 108 | ||
| BC006706.1 | - | 2 - 3 | 179 | 108 | ||
| BC014739.1 | - | 2 - 3 | 132 | 4920184 | 108 | |
| BC026569.1 | - | 2 - 3 | 132 | 4982612 | 108 | |
| BC029148.1 | - | 2 - 3 | 174 | 108 | ||
| BC029671.1 | - | 2 - 3 | 144 | 5364953 | 108 | |
| BC029694.1 | - | 2 - 3 | 141 | 5369477 | 108 | |
| BC033356.1 | - | 2 - 3 | 128 | 5366230 | 108 | |
| BG084120.2 | - | 2 - 3 | 201 | 108 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ik
Genomic Map