Assay Details
| Gene Symbol: Ik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011879.2 | NP_036009.1 | 9 - 10 | 922 | 89 | |
| GenBank mRNA | AJ006130.1 | - | 9 - 10 | 805 | 89 | |
| AK029644.1 | - | 9 - 10 | 876 | 89 | ||
| AK050625.1 | - | 9 - 10 | 880 | 89 | ||
| AK082356.1 | - | 9 - 10 | 854 | 89 | ||
| AK087976.1 | - | 9 - 10 | 868 | 89 | ||
| AK088908.1 | - | 8 - 9 | 1085 | 89 | ||
| AK163766.1 | - | 9 - 10 | 876 | 89 | ||
| AK167238.1 | - | 9 - 10 | 887 | 89 | ||
| BC006706.1 | - | 7 - 8 | 1280 | 89 | ||
| BC014739.1 | - | 9 - 10 | 853 | 4920184 | 89 | |
| BC026569.1 | - | 9 - 10 | 853 | 4982612 | 89 | |
| BC029148.1 | - | 9 - 10 | 895 | 89 | ||
| BC029671.1 | - | 9 - 10 | 865 | 5364953 | 89 | |
| BC029694.1 | - | 9 - 10 | 862 | 5369477 | 89 | |
| BC033356.1 | - | 9 - 10 | 849 | 5366230 | 89 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ik
Genomic Map