Assay Details
Gene Symbol: Tmco6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028036.3 | NP_082312.3 | 11 - 12 | 1472 | 71 | |
GenBank mRNA | AK010487.1 | - | 11 - 12 | 1397 | 71 | |
BC005637.1 | - | 11 - 12 | 1474 | 3156734 | 71 | |
BF468256.2 | - | 3 - 4 | 340 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625518 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625516 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625517 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292368 | BC005637.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292368 | BF468256.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292368 | NM_028036.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292368 | AK010487.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292368 | BC005637.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292368 | BF468256.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292368 | NM_028036.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292368 | AK010487.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 422339 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766500 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96011 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669401 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 302569 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 123786 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170165 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292368 | BC005637.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292368 | BF468256.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292368 | NM_028036.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292368 | AK010487.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292368 | BC005637.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292368 | BF468256.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292368 | NM_028036.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292368 | AK010487.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625569 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625519 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625558 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625546 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625562 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625549 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625520 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625526 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625557 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625551 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625553 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625527 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625555 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625566 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625559 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625561 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625544 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625529 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625516 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625568 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625523 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625521 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625518 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170165 | - | - |
Whole Mouse Genome G4122A | A_51_P292368 | BC005637.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P292368 | BF468256.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P292368 | NM_028036.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P292368 | AK010487.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P292368 | BC005637.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P292368 | BF468256.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P292368 | NM_028036.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P292368 | AK010487.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109225 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124626 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170165 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13887763 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13887757 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625516 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625517 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625558 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625608 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625566 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625553 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625489 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625578 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625617 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625544 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14117686 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625541 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625531 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625562 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625510 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13666518 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625520 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13777941 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625566 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13942419 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13666516 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625534 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625569 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625518 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625513 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625533 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625529 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625516 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625568 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625527 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625537 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625523 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625508 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13777940 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625549 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13969936 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625514 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625532 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14039649 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625551 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625526 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13887764 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14039650 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625540 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625543 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625521 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13777943 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31625546 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10454956 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454961 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454964 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454960 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454957 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454955 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454958 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454963 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454962 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454965 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454954 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454959 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451692_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1459587_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10454953 | - | - |
Mouse Genome 430 2.0 Array | 1459587_at | - | - |
Mouse Genome 430 2.0 Array | 1451692_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1451692_at | - | - |
Mouse Genome 430 2.0 Array | 1459587_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71983_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71983_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1800002617.mm.1 | NM_028036.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002617.mm.1 | NM_028036.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002617.mm.1 | TC1800000342.mm.1 | MTA TC |
Mouse Expression Set 430 | 1451692_at | - | - |
Mouse Expression Set 430 | 1451692_PM_at | - | - |
Mouse Expression Set 430 | 1459587_PM_at | - | - |
Mouse Expression Set 430 | 1459587_at | - | - |
Mouse Genome 430A 2.0 Array | 1451692_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1451692_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451692_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451692_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1459587_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1459587_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 420175 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2850348 | - | - |
MouseWG-6 v2 Expression BeadChip | 1990463 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2680487 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1990463 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2850348 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002680487 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002850348 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001990463 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2621853 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232025 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2620037 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2621853 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2620037 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1232025 | - | - |
mouse-6 v1.1 (Illumina) | scl0002151.1_1-S | - | - |
mouse-6 v1.1 (Illumina) | scl0002248.1_32-S | - | - |
mouse-6 v1.1 (Illumina) | scl52082.9.1_8-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2680487 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1990463 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2850348 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002151.1_1-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl52082.9.1_8-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002248.1_32-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2680487 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2850348 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1990463 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2621853 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2620037 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1232025 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2620037 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232025 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2621853 | - | - |
