Assay Details
| Gene Symbol: Ammecr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153515.4 | NP_705735.1 | 1 - 2 | 368 | 119 | |
| GenBank mRNA | AK054131.1 | - | 1 - 2 | 367 | 119 | |
| AK151060.1 | - | 1 - 2 | 369 | 119 | ||
| AK151257.1 | - | 1 - 2 | 269 | 119 | ||
| BC056948.1 | - | 1 - 2 | 312 | 6827318 | 119 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK151060.1, AK151257.1, BC056948.1, NM_153515.4 transcripts.
Set Membership: |
|

Ammecr1l
Genomic Map