Assay Details
| Gene Symbol: Prss58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175020.3 | NP_778185.1 | 3 - 4 | 339 | 76 | |
| GenBank mRNA | AK075754.1 | - | 3 - 4 | 340 | 76 | |
| BC048599.1 | - | 3 - 4 | 333 | 6740675 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prss58
Genomic Map