Assay Details
| Gene Symbol: Casd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145398.2 | NP_663373.2 | 11 - 12 | 1570 | 89 | |
| GenBank mRNA | AB091829.1 | - | 11 - 12 | 1570 | 89 | |
| BC018542.1 | - | 1 - 2 | 73 | 89 | ||
| BC033520.1 | - | 9 - 10 | 1194 | 89 | ||
| BC038009.1 | - | 1 - 2 | 96 | 89 | ||
| BC058953.1 | - | 3 - 4 | 563 | 89 | ||
| BC125377.1 | - | 11 - 12 | 1493 | 40129892 | 89 | |
| BC125379.1 | - | 11 - 12 | 1493 | 40129894 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC018542.1 transcripts.
Set Membership: |
|

Casd1
Genomic Map