Assay Details
| Gene Symbol: Casd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145398.2 | NP_663373.2 | 13 - 14 | 1814 | 79 | |
| GenBank mRNA | AB091829.1 | - | 13 - 14 | 1814 | 79 | |
| AK215801.1 | - | 3 - 4 | 254 | 79 | ||
| BC018542.1 | - | 3 - 4 | 317 | 79 | ||
| BC033520.1 | - | 11 - 12 | 1438 | 79 | ||
| BC038009.1 | - | 3 - 4 | 340 | 79 | ||
| BC058953.1 | - | 5 - 6 | 807 | 79 | ||
| BC125377.1 | - | 13 - 14 | 1737 | 40129892 | 79 | |
| BC125379.1 | - | 13 - 14 | 1737 | 40129894 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Casd1
Genomic Map