Assay Details
| Gene Symbol: Casd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145398.2 | NP_663373.2 | 15 - 16 | 2059 | 116 | |
| GenBank mRNA | AB091829.1 | - | 15 - 16 | 2059 | 116 | |
| BC018542.1 | - | 5 - 6 | 562 | 116 | ||
| BC033520.1 | - | 13 - 14 | 1683 | 116 | ||
| BC038009.1 | - | 5 - 6 | 585 | 116 | ||
| BC058953.1 | - | 7 - 8 | 1052 | 116 | ||
| BC125377.1 | - | 15 - 16 | 1982 | 40129892 | 116 | |
| BC125379.1 | - | 15 - 16 | 1982 | 40129894 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Casd1
Genomic Map