| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600000161.mm.1 |
TC0600000012.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000161.mm.1 |
NM_145398.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000161.mm.1 |
NM_145398.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000164.mm.1 |
TC0600000012.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000164.mm.1 |
NM_145398.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000164.mm.1 |
NM_145398.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000168.mm.1 |
TC0600000012.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000168.mm.1 |
NM_145398.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000168.mm.1 |
NM_145398.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000167.mm.1 |
TC0600000012.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000167.mm.1 |
NM_145398.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000167.mm.1 |
NM_145398.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000165.mm.1 |
TC0600000012.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000165.mm.1 |
NM_145398.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000165.mm.1 |
NM_145398.2 |
Refseq |