Assay Details
| Gene Symbol: Casd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145398.2 | NP_663373.2 | 4 - 5 | 498 | 135 | |
| GenBank mRNA | AB091829.1 | - | 4 - 5 | 498 | 135 | |
| BC033520.1 | - | 2 - 3 | 122 | 135 | ||
| BC125377.1 | - | 4 - 5 | 421 | 40129892 | 135 | |
| BC125379.1 | - | 4 - 5 | 421 | 40129894 | 135 | |
More Information
Updated Mapping Information
Set Membership: |
|

Casd1
Genomic Map