Assay Details
| Gene Symbol: Casd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145398.2 | NP_663373.2 | 9 - 10 | 1360 | 153 | |
| GenBank mRNA | AB091829.1 | - | 9 - 10 | 1360 | 153 | |
| BC033520.1 | - | 7 - 8 | 984 | 153 | ||
| BC058953.1 | - | 1 - 2 | 353 | 153 | ||
| BC125377.1 | - | 9 - 10 | 1283 | 40129892 | 153 | |
| BC125379.1 | - | 9 - 10 | 1283 | 40129894 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Casd1
Genomic Map