Assay Details
Gene Symbol: Sephs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009266.3 | NP_033292.2 | 1 | 1648 | 74 | |
GenBank mRNA | AK077573.1 | - | 1 | 1651 | 74 | |
AK149516.1 | - | 1 | 1645 | 74 | ||
AK168722.1 | - | 1 | 1649 | 74 | ||
AK168965.1 | - | 1 | 1649 | 74 | ||
BC016508.1 | - | 1 | 1640 | 4911695 | 74 | |
BC028966.1 | - | 1 | 1625 | 5374184 | 74 | |
BC043334.1 | - | 1 | 1629 | 5100207 | 74 | |
U43285.1 | - | 1 | 1645 | 74 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25504898 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25504897 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25504896 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25504894 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25504900 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25504895 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079324 | BC043334.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079324 | U43285.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079324 | BC016508.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079324 | BC028966.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079324 | AK149516.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596924 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130102 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079324 | BC043334.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079324 | U43285.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079324 | BC016508.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079324 | BC028966.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079324 | AK149516.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504889 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504923 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504927 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504893 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504904 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504925 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504896 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504913 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504877 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504905 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504885 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504901 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504903 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504898 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504899 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504921 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504892 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504919 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504886 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504888 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504909 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504910 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504907 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504908 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504879 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504915 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504922 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504882 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504890 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130102 | - | - |
Whole Mouse Genome G4122A | A_51_P418765 | BC043334.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P418765 | U43285.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P418765 | BC016508.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P418765 | BC028966.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P418765 | AK149516.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132839 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130102 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05907330 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05907328 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06066795 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06066797 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25504894 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25504900 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25504898 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25504896 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25504895 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25504897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504858 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504913 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504846 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504925 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504930 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504921 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504913 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504933 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06223069 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504890 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06011277 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504898 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504926 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504922 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504935 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06011275 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06457546 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504907 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05945318 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05907324 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06474371 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06612374 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06066797 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504910 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504875 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504888 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05907328 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504923 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504882 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06640182 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504904 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05907330 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504877 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504876 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504903 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06320804 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06222011 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504919 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504917 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504892 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504871 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05907336 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06222013 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25504925 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10568222 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10568223 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10568224 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418325_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449040_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10568221 | - | - |
Mouse Genome 430 2.0 Array | 1435602_at | - | - |
Mouse Genome 430 2.0 Array | 1449040_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1449040_a_at | - | - |
Mouse Genome 430 2.0 Array | 1418325_at | - | - |
Mouse Genome 430 2.0 Array | 1418325_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1435602_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20768_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20768_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700036292.mm.1 | NM_009266.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700036292.mm.1 | NM_009266.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700036292.mm.1 | TC0700004321.mm.1 | MTA TC |
Mouse Expression Set 430 | 1435602_PM_at | - | - |
Mouse Expression Set 430 | 1449040_PM_a_at | - | - |
Mouse Expression Set 430 | 1449040_a_at | - | - |
Mouse Expression Set 430 | 1418325_PM_at | - | - |
Mouse Expression Set 430 | 1435602_at | - | - |
Mouse Expression Set 430 | 1418325_at | - | - |
Murine Genome U74v2 Set | 93557_at | - | - |
Murine Genome U74v2 Set | 93557_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1449040_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1449040_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1435602_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418325_at | - | - |
Mouse Genome 430A 2.0 Array | 1435602_at | - | - |
Mouse Genome 430A 2.0 Array | 1418325_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1435602_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1435602_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418325_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449040_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418325_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449040_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6770446 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6770446 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006770446 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1217590 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1217590 | - | - |
mouse-6 v1.1 (Illumina) | scl30651.1_27-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6770446 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl30651.1_27-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6770446 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1217590 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1217590 | - | - |
