Assay Details
| Gene Symbol: Ace2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130513.1 | NP_001123985.1 | 2 - 3 | 517 | 140 | |
| NM_027286.4 | NP_081562.2 | 1 - 2 | 369 | 140 | ||
| GenBank mRNA | AB053181.1 | - | 1 - 2 | 373 | 140 | |
| AB053182.1 | - | 1 - 2 | 373 | 140 | ||
| AI115946.1 | - | 1 - 2 | 370 | 140 | ||
| AK141604.1 | - | 1 - 2 | 218 | 140 | ||
| BC026801.1 | - | 1 - 2 | 362 | 4236529 | 140 | |
| BY722909.1 | - | 2 - 3 | 518 | 140 | ||
| EF408739.1 | - | 2 - 3 | 478 | 140 | ||
| EF408740.1 | - | 2 - 3 | 478 | 140 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ace2
Genomic Map