Assay Details
| Gene Symbol: Spon1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145584.2 | NP_663559.1 | 15 - 16 | 2591 | 97 | |
| GenBank mRNA | AK146506.1 | - | 15 - 16 | 2415 | 97 | |
| AK173021.1 | - | 12 - 13 | 4126 | 97 | ||
| BC020531.1 | - | 15 - 16 | 2461 | 4221758 | 97 | |
| BC030339.1 | - | 6 - 7 | 1045 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spon1
Genomic Map