Assay Details
| Gene Symbol: Cct5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007637.2 | NP_031663.1 | 8 - 9 | 1282 | 70 | |
| GenBank mRNA | AK077753.1 | - | 8 - 9 | 1241 | 70 | |
| AK088184.1 | - | 8 - 9 | 1216 | 70 | ||
| AK129056.1 | - | 8 - 9 | 1188 | 70 | ||
| AK133857.1 | - | 8 - 9 | 1240 | 70 | ||
| AK136493.1 | - | 8 - 9 | 1234 | 70 | ||
| AK145445.1 | - | 8 - 9 | 1251 | 70 | ||
| AK146405.1 | - | 8 - 9 | 1254 | 70 | ||
| AK146425.1 | - | 8 - 9 | 1234 | 70 | ||
| AK150390.1 | - | 8 - 9 | 1201 | 70 | ||
| AK153911.1 | - | 8 - 9 | 1258 | 70 | ||
| AK167895.1 | - | 8 - 9 | 1223 | 70 | ||
| BC092233.1 | - | 8 - 9 | 1252 | 70 | ||
| BC094427.1 | - | 8 - 9 | 1229 | 30537966 | 70 | |
| Z31555.1 | - | 8 - 9 | 1282 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cct5
Genomic Map