Assay Details
Gene Symbol: Cdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016756.4 | NP_058036.1 | 2 - 3 | 418 | 125 | |
NM_183417.3 | NP_904326.1 | 2 - 3 | 418 | 125 | ||
GenBank mRNA | AJ223732.1 | - | 2 - 3 | 320 | 125 | |
AK042462.1 | - | 1 - 2 | 1079 | 125 | ||
AK146845.1 | - | 2 - 3 | 398 | 125 | ||
AK148017.1 | - | 2 - 3 | 357 | 125 | ||
AK151979.1 | - | 2 - 3 | 368 | 125 | ||
AK152922.1 | - | 2 - 3 | 371 | 125 | ||
AK153983.1 | - | 2 - 3 | 370 | 125 | ||
AK154997.1 | - | 2 - 3 | 370 | 125 | ||
AK181235.1 | - | 1 - 2 | 89 | 125 | ||
AK186022.1 | - | 1 - 2 | 89 | 125 | ||
AK189612.1 | - | 2 - 3 | 154 | 125 | ||
AK210959.1 | - | 1 - 2 | 125 | |||
BC005654.1 | - | 2 - 3 | 379 | 3709730 | 125 | |
BC026145.1 | - | 1 - 2 | 1124 | 125 | ||
CN705407.1 | - | 2 - 3 | 418 | 125 | ||
U63337.1 | - | 2 - 3 | 346 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
