Assay Details
| Gene Symbol: Sarnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025364.2 | NP_079640.1 | 7 - 8 | 565 | 105 | |
| GenBank mRNA | AK003453.1 | - | 7 - 8 | 417 | 105 | |
| AK018773.1 | - | 7 - 8 | 428 | 105 | ||
| AK088266.1 | - | 7 - 8 | 414 | 105 | ||
| AK146248.1 | - | 7 - 8 | 443 | 105 | ||
| BC027510.1 | - | 7 - 8 | 404 | 1382354 | 105 | |
| BC157909.1 | - | 7 - 8 | 429 | 9088111 | 105 | |
| BC158023.1 | - | 7 - 8 | 429 | 9007329 | 105 | |
| BC172063.1 | - | 7 - 8 | 426 | 9054349 | 105 | |
| BF148013.1 | - | 4 - 5 | 105 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sarnp
Genomic Map