Assay Details
Gene Symbol: Suox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173733.3 | NP_776094.2 | 1 - 2 | 71 | 69 | |
GenBank mRNA | AK149422.1 | - | 1 - 2 | 59 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK149422.1, NM_173733.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
