Assay Details
| Gene Symbol: Scgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145399.1 | NP_663374.1 | 8 - 9 | 751 | 116 | |
| GenBank mRNA | BC016093.1 | - | 8 - 9 | 751 | 4504330 | 116 |
More Information
Set Membership: |
|

Scgn
Genomic Map