Assay Details
| Gene Symbol: Tas2r125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207027.1 | NP_996910.1 | 1 | 759 | 156 | |
| GenBank mRNA | BC107190.1 | - | 1 | 759 | 40050840 | 156 |
| BC107191.1 | - | 1 | 759 | 40050841 | 156 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069197 | NM_207027.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069197 | BC107190.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069197 | BC107191.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 30328 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 47637 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131660 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069197 | NM_207027.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069197 | BC107190.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069197 | BC107191.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24855175 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131660 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131660 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05608243 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05608244 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05608252 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05608262 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05608257 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05608265 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05608242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24855155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05669292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05040528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24855177 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10542258 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10542257 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600012624.mm.1 | TC0600001568.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600012624.mm.1 | NM_207027.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012624.mm.1 | NM_207027.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 406438 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6550600 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6550600 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006550600 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213409 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213409 | - | - |
| mouse-6 v1.1 (Illumina) | scl0387352.1_137-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6550600 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0387352.1_137-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6550600 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1213409 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213409 | - | - |

Tas2r125
Genomic Map