Assay Details
| Gene Symbol: Tac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199971.1 | NP_001186900.1 | 7 - 8 | 630 | 86 | |
| NM_009312.2 | NP_033338.2 | 6 - 7 | 458 | 86 | ||
| GenBank mRNA | AF186116.1 | - | 6 - 7 | 424 | 86 | |
| BC031348.1 | - | 6 - 7 | 455 | 86 | ||
| BC051476.1 | - | 6 - 7 | 464 | 1533442 | 86 | |
| BQ175657.1 | - | 1 - 2 | 86 | |||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2147566 | BQ175657.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2147566 | D14423.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2147566 | NM_001199971.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2147566 | NM_009312.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2147566 | AF186116.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528251 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 194724 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 259432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737810 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 856057 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 157490 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102134 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2147566 | BQ175657.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2147566 | D14423.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2147566 | NM_001199971.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2147566 | NM_009312.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2147566 | AF186116.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P415087 | D14423.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537722 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537720 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537748 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537719 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537755 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02246252 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102134 | - | - |
| Whole Mouse Genome G4122A | A_51_P415087 | D14423.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116054 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102134 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08514403 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08482517 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08559264 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08514472 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08559262 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08559263 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08482509 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08908916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08514451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08845670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08597462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08424770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08845671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08908916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08514449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08845673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08349704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08514409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08424775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08514464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08514472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537732 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10367031 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367030 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367029 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367032 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367027 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367028 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367026 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367025 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419411_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10367024 | - | - |
| Mouse Genome 430 2.0 Array | 1419411_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419411_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21334_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21334_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa1300_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000011305.mm.1 | NM_001199971.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011305.mm.1 | NM_001199971.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011305.mm.1 | NM_009312.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011305.mm.1 | NM_009312.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011305.mm.1 | TC1000001571.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011309.mm.1 | NM_001199971.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011309.mm.1 | NM_001199971.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011309.mm.1 | NM_009312.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011309.mm.1 | NM_009312.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011309.mm.1 | TC1000001571.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011311.mm.1 | NM_001199971.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011311.mm.1 | NM_001199971.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011311.mm.1 | NM_009312.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011311.mm.1 | NM_009312.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011311.mm.1 | TC1000001571.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011304.mm.1 | NM_009312.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011304.mm.1 | NM_009312.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011304.mm.1 | TC1000001571.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011310.mm.1 | NM_001199971.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011310.mm.1 | NM_001199971.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011310.mm.1 | NM_009312.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011310.mm.1 | NM_009312.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011310.mm.1 | TC1000001571.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011306.mm.1 | NM_001199971.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011306.mm.1 | NM_001199971.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011306.mm.1 | NM_009312.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011306.mm.1 | NM_009312.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011306.mm.1 | TC1000001571.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011302.mm.1 | NM_001199971.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011302.mm.1 | NM_001199971.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011302.mm.1 | TC1000001571.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011308.mm.1 | NM_001199971.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011308.mm.1 | NM_001199971.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011308.mm.1 | NM_009312.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011308.mm.1 | NM_009312.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011308.mm.1 | TC1000001571.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011307.mm.1 | NM_001199971.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011307.mm.1 | NM_001199971.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011307.mm.1 | NM_009312.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011307.mm.1 | NM_009312.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011307.mm.1 | TC1000001571.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419411_PM_at | - | - |
| Mouse Expression Set 430 | 1419411_at | - | - |
| Murine Genome U74v2 Set | 103548_at | - | - |
| Murine Genome U74v2 Set | 103548_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419411_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419411_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419411_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419411_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 380706 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000380706 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2771036 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2771036 | - | - |
| mouse-6 v1.1 (Illumina) | scl38312.8.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 380706 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38312.8.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380706 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2790810 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2790810 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2771036 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl38312.8.1_1-S | - | - |

Tac2
Genomic Map