Assay Details
| Gene Symbol: Tmem194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113211.1 | NP_001106682.1 | 7 - 8 | 1006 | 134 | |
| GenBank mRNA | AK129110.1 | - | 7 - 8 | 1003 | 134 | |
| AK132974.1 | - | 1475 | 134 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem194
Genomic Map