Assay Details
| Gene Symbol: Atp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016774.3 | NP_058054.2 | 2 - 3 | 332 | 75 | |
| GenBank mRNA | AK002621.1 | - | 2 - 3 | 329 | 75 | |
| AK010314.1 | - | 2 - 3 | 332 | 75 | ||
| AK011035.1 | - | 2 - 3 | 332 | 75 | ||
| AK083276.1 | - | 2 - 3 | 334 | 75 | ||
| AK084009.1 | - | 2 - 3 | 323 | 75 | ||
| AK145684.1 | - | 2 - 3 | 333 | 75 | ||
| AK148891.1 | - | 2 - 3 | 334 | 75 | ||
| AK150599.1 | - | 2 - 3 | 334 | 75 | ||
| AK151081.1 | - | 2 - 3 | 334 | 75 | ||
| AK151600.1 | - | 2 - 3 | 331 | 75 | ||
| AK152788.1 | - | 2 - 3 | 334 | 75 | ||
| AK152976.1 | - | 2 - 3 | 334 | 75 | ||
| AK153099.1 | - | 2 - 3 | 323 | 75 | ||
| AK159444.1 | - | 2 - 3 | 334 | 75 | ||
| AK159737.1 | - | 2 - 3 | 337 | 75 | ||
| AK159978.1 | - | 2 - 3 | 334 | 75 | ||
| AK160199.1 | - | 2 - 3 | 326 | 75 | ||
| AK160608.1 | - | 2 - 3 | 334 | 75 | ||
| AK164383.1 | - | 2 - 3 | 322 | 75 | ||
| AK166525.1 | - | 2 - 3 | 335 | 75 | ||
| AK166603.1 | - | 2 - 3 | 334 | 75 | ||
| AK166979.1 | - | 2 - 3 | 333 | 75 | ||
| AK167119.1 | - | 2 - 3 | 334 | 75 | ||
| AK167160.1 | - | 2 - 3 | 324 | 75 | ||
| AK167728.1 | - | 2 - 3 | 331 | 75 | ||
| AK167764.1 | - | 2 - 3 | 334 | 75 | ||
| AK168692.1 | - | 2 - 3 | 324 | 75 | ||
| AK168941.1 | - | 2 - 3 | 334 | 75 | ||
| AK169184.1 | - | 2 - 3 | 334 | 75 | ||
| AK195546.1 | - | 1 - 2 | 97 | 75 | ||
| BC013253.1 | - | 2 - 3 | 308 | 75 | ||
| BC037127.1 | - | 2 - 3 | 325 | 75 | ||
| BC046616.1 | - | 2 - 3 | 315 | 6494223 | 75 | |
| DQ403100.1 | - | 1 - 2 | 120 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp5b
Genomic Map