Assay Details
| Gene Symbol: Krt28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027574.1 | NP_081850.1 | 7 - 8 | 1306 | 87 | |
| GenBank mRNA | AB288231.1 | - | 7 - 8 | 1306 | 87 | |
| AK014642.1 | - | 7 - 8 | 1307 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976972 | NM_027574.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976972 | AB288231.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976972 | AK014642.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964672 | NM_027574.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964672 | AB288231.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964672 | AK014642.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930933 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 142082 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 214399 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 403388 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 52298 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 63618 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142270 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114431 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976972 | NM_027574.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976972 | AB288231.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976972 | AK014642.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964672 | NM_027574.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964672 | AB288231.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964672 | AK014642.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114431 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142270 | - | - |
| Whole Mouse Genome G4122A | A_51_P488780 | AK014642.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142270 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09527741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09569024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09714806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09527739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09464567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061901 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09156941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061897 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061883 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10390828 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390826 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390830 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390824 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390829 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390825 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390827 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430132_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458354_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10390823 | - | - |
| Mouse Genome 430 2.0 Array | 1430132_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458354_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1430132_at | - | - |
| Mouse Genome 430 2.0 Array | 1458354_x_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100034185.mm.1 | NM_027574.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034185.mm.1 | NM_027574.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034185.mm.1 | TC1100003705.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430132_at | - | - |
| Mouse Expression Set 430 | 1458354_x_at | - | - |
| Mouse Expression Set 430 | 1458354_PM_x_at | - | - |
| Mouse Expression Set 430 | 1430132_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430132_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458354_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430132_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458354_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2624267 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2624267 | - | - |

Krt28
Genomic Map