Assay Details
Gene Symbol: Olfr771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146547.1 | NP_666758.1 | 1 | 535 | 101 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057751 | NM_146547.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104567 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145591 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170830 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057751 | NM_146547.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P479138 | NM_146547.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544934 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544936 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544941 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544953 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544933 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544965 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544951 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544931 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544932 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544927 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544968 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544945 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544929 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544954 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544923 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544947 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544930 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544962 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544960 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544938 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544958 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544940 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544928 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544943 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544946 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544966 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544956 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544964 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544949 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544944 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104567 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145591 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170830 | - | - |
Whole Mouse Genome G4122A | A_51_P479138 | NM_146547.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127433 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170830 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145591 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104567 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08943391 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08943396 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08943385 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08943378 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08943397 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08943390 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08943382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544909 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544966 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544967 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544938 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544907 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544971 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544921 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544920 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544967 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544962 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08946670 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544940 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544915 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544954 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544931 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544981 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544966 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08425310 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544977 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544918 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544969 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544974 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544917 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544906 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544919 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544970 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08724959 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544928 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544930 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544905 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544911 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544945 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544909 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544956 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544934 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544947 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544943 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544936 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544964 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544960 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08926787 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544958 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544923 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08943387 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08569920 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544933 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08926788 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544951 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544973 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27544932 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10373619 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10373618 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 810680 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 3450064 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003450064 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2697776 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2697776 | - | - |
mouse-6 v1.1 (Illumina) | scl5164.1.1_48-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3450064 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl5164.1.1_48-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3450064 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2901331 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2901331 | - | - |
