Assay Details
| Gene Symbol: Agpat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018862.3 | NP_061350.3 | 1 - 2 | 65 | 56 | |
| GenBank mRNA | BC009651.1 | - | 1 - 2 | 65 | 3487719 | 56 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC009651.1, NM_018862.3 transcripts.
Set Membership: |
|

Agpat1
Genomic Map