Assay Details
Gene Symbol: Gpm6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177957.1 | NP_001171428.1 | 8 - 9 | 1397 | 87 | |
NM_001177960.1 | NP_001171431.1 | 7 - 8 | 1277 | 87 | ||
NM_001177962.1 | NP_001171433.1 | 7 - 8 | 942 | 87 | ||
GenBank mRNA | AF254872.1 | - | 7 - 8 | 1138 | 87 | |
AF254874.1 | - | 8 - 9 | 1258 | 87 | ||
AF254876.1 | - | 7 - 8 | 913 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
