Assay Details
| Gene Symbol: Iqub | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172535.3 | NP_766123.2 | 10 - 11 | 1844 | 142 | |
| GenBank mRNA | AK029931.1 | - | 10 - 11 | 1834 | 142 | |
| AK133019.1 | - | 10 - 11 | 1826 | 142 | ||
| AK161501.1 | - | 4 - 5 | 669 | 142 | ||
| BC137937.1 | - | 10 - 11 | 1811 | 8860956 | 142 | |
| BC137957.1 | - | 10 - 11 | 1811 | 8860976 | 142 | |
More Information
Set Membership: |
|

Iqub
Genomic Map