Assay Details
| Gene Symbol: Asz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023729.3 | NP_076218.3 | 8 - 9 | 904 | 140 | |
| GenBank mRNA | AF459789.1 | - | 8 - 9 | 898 | 140 | |
| BC058859.1 | - | 8 - 9 | 913 | 30011651 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asz1
Genomic Map