Assay Details
Gene Symbol: Gmnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020567.2 | NP_065592.1 | 1 - 2 | 119 | 64 | |
GenBank mRNA | AF068780.1 | - | 266 | 64 | ||
AK131959.1 | - | 1 - 2 | 93 | 64 | ||
BY045588.1 | - | 1 - 2 | 119 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF068780.1, AK131959.1, NM_020567.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
