Assay Details
Gene Symbol: Smo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176996.4 | NP_795970.3 | 11 - 12 | 2459 | 76 | |
GenBank mRNA | AK081050.1 | - | 5 - 6 | 647 | 76 | |
BC030377.1 | - | 4 - 5 | 496 | 76 | ||
BC096028.1 | - | 11 - 12 | 1978 | 6850065 | 76 | |
FJ883552.1 | - | 11 - 12 | 1968 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
