Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600001655.mm.1 |
TC0600000242.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600001655.mm.1 |
NM_176996.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001655.mm.1 |
NM_176996.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001651.mm.1 |
TC0600000242.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600001651.mm.1 |
NM_176996.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001651.mm.1 |
NM_176996.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001652.mm.1 |
TC0600000242.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600001652.mm.1 |
NM_176996.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001652.mm.1 |
NM_176996.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001653.mm.1 |
TC0600000242.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600001653.mm.1 |
NM_176996.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001653.mm.1 |
NM_176996.4 |
Refseq |