Assay Details
Gene Symbol: Galnt18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173739.3 | NP_776100.2 | 1 - 2 | 661 | 92 | |
GenBank mRNA | AB175685.1 | - | 1 - 2 | 231 | 92 | |
BC024988.1 | - | 1 - 2 | 661 | 5044626 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
