Assay Details
| Gene Symbol: Rheb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053075.3 | NP_444305.2 | 5 - 6 | 457 | 80 | |
| GenBank mRNA | AB039919.1 | - | 5 - 6 | 507 | 80 | |
| AK151213.1 | - | 5 - 6 | 489 | 80 | ||
| AY197373.1 | - | 5 - 6 | 457 | 80 | ||
| BC012273.1 | - | 5 - 6 | 474 | 3710454 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rheb
Genomic Map