Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700014527.mm.1 |
TC0700001726.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014527.mm.1 |
NM_177213.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014527.mm.1 |
NM_177213.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014526.mm.1 |
TC0700001726.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014526.mm.1 |
NM_177213.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014526.mm.1 |
NM_177213.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014528.mm.1 |
TC0700001726.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014528.mm.1 |
NM_177213.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014528.mm.1 |
NM_177213.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014525.mm.1 |
TC0700001726.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014525.mm.1 |
NM_177213.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014525.mm.1 |
NM_177213.3 |
Refseq |