Assay Details
Gene Symbol: Tpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK136486.1 | - | 6 - 7 | 412 | 75 | |
BC023354.1 | - | 6 - 7 | 387 | 5253545 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
