Assay Details
| Gene Symbol: Sec23ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029982.2 | NP_001025153.2 | 12 - 13 | 2258 | 82 | |
| GenBank mRNA | BC039781.1 | - | 4 - 5 | 442 | 82 | |
| BC066195.1 | - | 11 - 12 | 1889 | 82 | ||
| BC098506.1 | - | 12 - 13 | 2147 | 30014037 | 82 | |
| BC116304.1 | - | 11 - 12 | 1639 | 82 | ||
| BC116305.1 | - | 11 - 12 | 1639 | 82 | ||
More Information
Set Membership: |
|

Sec23ip
Genomic Map