Assay Details
| Gene Symbol: Sec23ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029982.2 | NP_001025153.2 | 17 - 18 | 3065 | 130 | |
| GenBank mRNA | AK042851.1 | - | 3 - 4 | 418 | 130 | |
| AK134922.1 | - | 3 - 4 | 466 | 130 | ||
| BC023480.1 | - | 4 - 5 | 511 | 130 | ||
| BC039781.1 | - | 9 - 10 | 1182 | 130 | ||
| BC098506.1 | - | 17 - 18 | 2954 | 30014037 | 130 | |
| BC116304.1 | - | 16 - 17 | 2446 | 130 | ||
| BC116305.1 | - | 16 - 17 | 2446 | 130 | ||
More Information
Set Membership: |
|

Sec23ip
Genomic Map