Assay Details
| Gene Symbol: Sec23ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029982.2 | NP_001025153.2 | 7 - 8 | 1545 | 71 | |
| GenBank mRNA | AK048053.1 | - | 7 - 8 | 1474 | 71 | |
| BC066195.1 | - | 6 - 7 | 1176 | 71 | ||
| BC098506.1 | - | 7 - 8 | 1434 | 30014037 | 71 | |
| BC116304.1 | - | 6 - 7 | 926 | 71 | ||
| BC116305.1 | - | 6 - 7 | 926 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec23ip
Genomic Map