Assay Details
Gene Symbol: Prl8a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023332.3 | NP_075821.1 | 4 - 5 | 413 | 114 | |
GenBank mRNA | AF158744.1 | - | 4 - 5 | 398 | 114 | |
AK005409.1 | - | 4 - 5 | 414 | 114 | ||
AK005494.1 | - | 4 - 5 | 414 | 114 | ||
BC064762.1 | - | 4 - 5 | 381 | 30013931 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509999 | NM_023332.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509999 | AF158744.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509999 | AK005409.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509999 | AK005494.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509999 | BC064762.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509999 | NM_023332.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509999 | AF158744.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509999 | AK005409.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509999 | AK005494.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509999 | BC064762.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 462383 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251238 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 614293 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340847 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 520747 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171295 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509999 | NM_023332.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509999 | AF158744.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509999 | AK005409.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509999 | AK005494.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509999 | BC064762.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509999 | NM_023332.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509999 | AF158744.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509999 | AK005409.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509999 | AK005494.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509999 | BC064762.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902674 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902679 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902678 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902655 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902651 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902648 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902637 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902647 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902646 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902669 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902664 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902659 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902676 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902682 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902671 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902652 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902681 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902642 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902665 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902635 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902640 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902684 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902667 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902653 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902668 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902641 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902638 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902658 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902675 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902634 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902661 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902666 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902657 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02652222 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171295 | - | - |
Whole Mouse Genome G4122A | A_51_P509999 | NM_023332.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P509999 | AF158744.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P509999 | AK005409.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P509999 | AK005494.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P509999 | BC064762.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P509999 | NM_023332.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P509999 | AF158744.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P509999 | AK005409.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P509999 | AK005494.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P509999 | BC064762.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138960 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129086 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171295 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11099447 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11099445 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11099433 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11099448 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11099432 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11099438 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11099444 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902608 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902620 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902658 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902608 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902655 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902681 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902611 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902671 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902684 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902648 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902653 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902620 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902609 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902682 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902642 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902606 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902626 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902637 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902651 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902627 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902657 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902598 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902618 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902686 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902640 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902599 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902628 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902635 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902624 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902679 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902612 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902623 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902634 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902629 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902663 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902664 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902633 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902638 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902669 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902602 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902621 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902600 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902605 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902614 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902665 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902644 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902661 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902616 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902619 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902674 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902604 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902667 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902676 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902675 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11099444 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902668 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902678 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902631 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902666 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10711174 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10408398 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408396 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408401 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408400 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408399 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408397 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1448532_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10408395 | - | - |
Mouse Genome 430 2.0 Array | 1448532_at | - | - |
Mouse Genome 430 2.0 Array | 1448532_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67310_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67310_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300010521.mm.1 | NM_023332.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010521.mm.1 | TC1300001692.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300010522.mm.1 | NM_023332.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010522.mm.1 | TC1300001692.mm.1 | MTA TC |
Mouse Expression Set 430 | 1448532_at | - | - |
Mouse Expression Set 430 | 1448532_PM_at | - | - |
Murine Genome U74v2 Set | 169543_f_at | - | - |
Murine Genome U74v2 Set | 169543_PM_f_at | - | - |
Mouse Genome 430A 2.0 Array | 1448532_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1448532_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448532_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448532_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 302539 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4060156 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004060156 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1256562 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1256562 | - | - |
mouse-6 v1.1 (Illumina) | scl44170.6.1_52-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4060156 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl44170.6.1_52-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4060156 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1256562 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1256562 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl44170.6.1_52-S | - | - |
